ContactUsLinkComponent
PAHs Mixture - 23 components: 1000ug/ml each of Naphthalene [CAS:91-20-3] Acenaphthylene [CAS:208-96-8] Acenaphthene [CAS:83-32-9] Fluorene [CAS:86-73-7] Phenanthrene [CAS:85-01-8] Anthracene [CAS:120-12-7] Fluoranthene [CAS:206-44-0] P 1 * 1 mL
Catalog # ACSDF223355
Supplier:  CPACHEM
New Product
PAHs Mixture - 23 components: 1000ug/ml each of Naphthalene [CAS:91-20-3] Acenaphthylene [CAS:208-96-8] Acenaphthene [CAS:83-32-9] Fluorene [CAS:86-73-7] Phenanthrene [CAS:85-01-8] Anthracene [CAS:120-12-7] Fluoranthene [CAS:206-44-0] P 1 * 1 mL
Catalog # ACSDF223355
Supplier:  CPACHEM