ContactUsLinkComponent
Fixed high intensity multiplexing LEDs with peak band passes of Red (623nm), Green (5205m), Blue (465nm), Infra-Red (740nm), Far-Red (638nm) 1 * 1 items
Catalog # SYNOHILED-RGBIRFR
New Product
Fixed high intensity multiplexing LEDs with peak band passes of Red (623nm), Green (5205m), Blue (465nm), Infra-Red (740nm), Far-Red (638nm) 1 * 1 items
Catalog # SYNOHILED-RGBIRFR