1/16 GC ferrule 0.3 mm ID (long) three-hole 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215193
Supplier: Trajan Scientific and Medical
New Product
1/16 GC ferrule 0.3 mm ID (long) three-hole 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215193
Supplier: Trajan Scientific and Medical
New Product
Frequently Bought Together
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Certified screw top vials and closures
Supplier: Agilent
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Inlet septa
Supplier: Agilent
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
Total organic carbon (TOC) vials
Supplier: EP SCIENTIFIC
Microliter™ 700 series syringes
Supplier: HAMILTON BONADUZ
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Exhaust filters, b.safe
Supplier: Bohlender
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Inserts for Screw Neck Vials ND8
Supplier: VWR Collection
Glass screw top 0,3 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
Screw caps for certified screw top vials
Supplier: Agilent