N25/500 1.5BV NEEDLES PK5 1 * 5 items
Catalog # SGEA039170
Supplier: Trajan Scientific and Medical
New Product
N25/500 1.5BV NEEDLES PK5 1 * 5 items
Catalog # SGEA039170
Supplier: Trajan Scientific and Medical
New Product
Frequently Bought Together
HPLC columns, Allure® silica (USP L3)
Supplier: Restek
ISAspher HPLC Columns, C18 AQ
Supplier: ISERA
Metab-AAC HPLC Columns, BF Series
Supplier: ISERA
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
COLUMN 130Å 2.5 µM C18 XBRIDGE BEH XP 1 * 1 items
Supplier: WATERS
ISAspher HPLC Columns, C8 Select
Supplier: ISERA
Discovery® Classical C18 HPLC Columns
Supplier: Merck
Safety caps and safety waste cap accessories and replacement parts
Supplier: S.C.A.T.
Size exclusion chromatography columns, Bio SEC-3 and Bio SEC-5
Supplier: Agilent
VWR®, Screw Neck Vials ND10
Supplier: VWR Collection
PEPMAP100 C18 3UM 75UMX2CM NVTRAP 1200 1 * 1 items
Supplier: Thermo Fisher Scientific
Adapter ferrules, Valco®
Supplier: Restek
Crimp Caps for Crimp Neck Vials, N 11
Supplier: MACHEREY-NAGEL