ContactUsLinkComponent
Muffle Furnace LT9/14/B510 Tmax1400 with lift door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3379
Supplier:  NABERTHERM
New Product
Muffle Furnace LT9/14/B510 Tmax1400 with lift door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3379
Supplier:  NABERTHERM