ContactUsLinkComponent
Muffle Furnace L5/14/B510 Tmax1400 with flap door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3312
Supplier:  NABERTHERM
New Product
Muffle Furnace L5/14/B510 Tmax1400 with flap door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3312
Supplier:  NABERTHERM