ContactUsLinkComponent
Muffle Furnace L9/11/B510 Tmax1100 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Catalog # 455-2718
Supplier:  NABERTHERM
New Product
Muffle Furnace L9/11/B510 Tmax1100 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Catalog # 455-2718
Supplier:  NABERTHERM