MALONATE STANDARD SOLUTION 1000 µG/ML FO 1 * 100 mL
Catalog # CHMBCL01.1384.0100
Supplier: CHEMLAB
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions, ready to use
Supplier: Bernd Kraft
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
TROLAMINE IMPURITY A ETHANOLAMINE 1 * 250 mg
Supplier: EDQM
DIISOPROPYL ETHER 1 * 3 items
Supplier: Merck
SIMETRYN 1 * 100 mg
Supplier: Merck
VWR® ARISTAR®, Cadmium Standard Solutions for ICP-MS
Supplier: VWR Chemicals
Glucose standard for IC, TraceCERT®, Supelco®
Supplier: Merck
ARISTAR®VWR® , Silicon Standard Solutions for ICP-MS
Supplier: VWR Chemicals