ContactUsLinkComponent
Electrical sterilization system SteriMax UK PLUG with 7 angular adjustments, residual heat display, tray for 3 instruments - Stainless steel design - Long-lasting sterilization tube Made of quartz glass - Sterilization period 5-7 seconds - Self-locki 1 * 1 items
Catalog # WART5.001.000_UK
Supplier:  WARTEWIG
New Product
Electrical sterilization system SteriMax UK PLUG with 7 angular adjustments, residual heat display, tray for 3 instruments - Stainless steel design - Long-lasting sterilization tube Made of quartz glass - Sterilization period 5-7 seconds - Self-locki 1 * 1 items
Catalog # WART5.001.000_UK
Supplier:  WARTEWIG