ContactUsLinkComponent
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 2 x 9 1 * 1.000 items
Catalog # KACC00209TWBBSSSD
New Product
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 2 x 9 1 * 1.000 items
Catalog # KACC00209TWBBSSSD