ContactUsLinkComponent
Stopper Bag, Tyvek & HDPE 28 x 40 , with Steam Sterilization Indicator 1 * 125 items
Catalog # KACC2840SBSD
New Product
Stopper Bag, Tyvek & HDPE 28 x 40 , with Steam Sterilization Indicator 1 * 125 items
Catalog # KACC2840SBSD