ContactUsLinkComponent
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 6 x 35 1 * 500 items
Catalog # KACC00635TWBBSSSD
New Product
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 6 x 35 1 * 500 items
Catalog # KACC00635TWBBSSSD