ContactUsLinkComponent
Human Amyloid beta Peptide 1-40 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quan titative determination of human Amyloid beta Peptide 1-40 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1802-96
Supplier:  ANTIBODIES.COM
New Product
Human Amyloid beta Peptide 1-40 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quan titative determination of human Amyloid beta Peptide 1-40 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1802-96
Supplier:  ANTIBODIES.COM