ContactUsLinkComponent
Human Pancreatic alpha Amylase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Pancreatic alpha Amylase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270640-48
Supplier:  ANTIBODIES.COM
New Product
Human Pancreatic alpha Amylase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Pancreatic alpha Amylase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270640-48
Supplier:  ANTIBODIES.COM