ContactUsLinkComponent
Human Amphiregulin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Amphiregulin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 48 Tests
Catalog # ANTIA1648-48
Supplier:  ANTIBODIES.COM
New Product
Human Amphiregulin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Amphiregulin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 48 Tests
Catalog # ANTIA1648-48
Supplier:  ANTIBODIES.COM