ContactUsLinkComponent
Human Heme Oxygenase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Heme Oxygenase 1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 96 Tests
Catalog # ANTIA1785-96
Supplier:  ANTIBODIES.COM
New Product
Human Heme Oxygenase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Heme Oxygenase 1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 96 Tests
Catalog # ANTIA1785-96
Supplier:  ANTIBODIES.COM