ContactUsLinkComponent
Mouse Cannabinoid Receptor II ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse Cannabinoid Receptor II in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270879-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Cannabinoid Receptor II ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse Cannabinoid Receptor II in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270879-48
Supplier:  ANTIBODIES.COM