ContactUsLinkComponent
Human LGALS3BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human LGALS3BP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA2828-48
Supplier:  ANTIBODIES.COM
New Product
Human LGALS3BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human LGALS3BP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA2828-48
Supplier:  ANTIBODIES.COM