ContactUsLinkComponent
Human Thromboxane A2 Receptor / TBXA2R ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Thromboxane A2 Receptor / TBXA2R in serum, plasma, and other biological 1 * 96 Tests
  ANTIA312698-96
New Product
Human Thromboxane A2 Receptor / TBXA2R ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Thromboxane A2 Receptor / TBXA2R in serum, plasma, and other biological 1 * 96 Tests
  ANTIA312698-96