ContactUsLinkComponent
Human Borrelia + Tick-Borne Encephalitis Virus IgG LIA is a Line Immunoassay designed for the in vitro qualititative determination o f human Borrelia + TBEV IgG in serum, plasma, cerebrospinal fluid, and synovial fluid. 1 * 16 Tests
Catalog # ANTIA324370-16
Supplier:  ANTIBODIES.COM
New Product
Human Borrelia + Tick-Borne Encephalitis Virus IgG LIA is a Line Immunoassay designed for the in vitro qualititative determination o f human Borrelia + TBEV IgG in serum, plasma, cerebrospinal fluid, and synovial fluid. 1 * 16 Tests
Catalog # ANTIA324370-16
Supplier:  ANTIBODIES.COM