ContactUsLinkComponent
Mouse Tryptophan Hydroxylase/TPH ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of mouse Tryptophan Hydroxylase/TPH in serum, plasma, cell culture supernatant, cell or tissue l 1 * 96 Tests
Catalog # ANTIA326931-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Tryptophan Hydroxylase/TPH ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of mouse Tryptophan Hydroxylase/TPH in serum, plasma, cell culture supernatant, cell or tissue l 1 * 96 Tests
Catalog # ANTIA326931-96
Supplier:  ANTIBODIES.COM