ContactUsLinkComponent
Mouse PSAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse PSAP in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalog # ANTIA326846-96
Supplier:  ANTIBODIES.COM
New Product
Mouse PSAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse PSAP in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalog # ANTIA326846-96
Supplier:  ANTIBODIES.COM