ContactUsLinkComponent
Porcine Terminal Complement Complex C5b-9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of porcine Terminal Complement Complex C5b-9 in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA319490-96
Supplier:  ANTIBODIES.COM
New Product
Porcine Terminal Complement Complex C5b-9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of porcine Terminal Complement Complex C5b-9 in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA319490-96
Supplier:  ANTIBODIES.COM