ContactUsLinkComponent
Human Borrelia IgM burgdorferi sensu stricto LIA is a Line Immunoassay designed for the in vitro qualititative determination of huma n Borrelia IgM burgdorferi sensu stricto in serum, plasma, cerebrospinal fluid, and synovial fluid. 1 * 16 Tests
Catalog # ANTIA324378-16
Supplier:  ANTIBODIES.COM
New Product
Human Borrelia IgM burgdorferi sensu stricto LIA is a Line Immunoassay designed for the in vitro qualititative determination of huma n Borrelia IgM burgdorferi sensu stricto in serum, plasma, cerebrospinal fluid, and synovial fluid. 1 * 16 Tests
Catalog # ANTIA324378-16
Supplier:  ANTIBODIES.COM