ContactUsLinkComponent
Human Glutathione Peroxidase 2 / GPX2 ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Glutathione Peroxidase 2 / GPX2 in serum, plasma, and other biological fl 1 * 96 Tests
Catalog # ANTIA313804-96
Supplier:  ANTIBODIES.COM
New Product
Human Glutathione Peroxidase 2 / GPX2 ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Glutathione Peroxidase 2 / GPX2 in serum, plasma, and other biological fl 1 * 96 Tests
Catalog # ANTIA313804-96
Supplier:  ANTIBODIES.COM