ContactUsLinkComponent
Mouse CD62P ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse CD62P in serum, platelet-poor plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 96 Tests
  ANTIA3165-96
New Product
Mouse CD62P ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse CD62P in serum, platelet-poor plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 96 Tests
  ANTIA3165-96