ContactUsLinkComponent
Human Acetyl Coenzyme A Carboxylase alpha ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Acetyl Coenzyme A Carboxylase alpha in serum, plasma, and other biolo 1 * 96 Tests
Catalog # ANTIA310746-96
Supplier:  ANTIBODIES.COM
New Product
Human Acetyl Coenzyme A Carboxylase alpha ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Acetyl Coenzyme A Carboxylase alpha in serum, plasma, and other biolo 1 * 96 Tests
Catalog # ANTIA310746-96
Supplier:  ANTIBODIES.COM