ContactUsLinkComponent
Human C-Peptide ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of human C-Peptide in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 96 Tests
Catalog # ANTIA3155-96
Supplier:  ANTIBODIES.COM
New Product
Human C-Peptide ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of human C-Peptide in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 96 Tests
Catalog # ANTIA3155-96
Supplier:  ANTIBODIES.COM