ContactUsLinkComponent
Monkey Plasminogen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of monkey Plasminogen in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samp 1 * 96 Tests
Catalog # ANTIA326834-96
Supplier:  ANTIBODIES.COM
New Product
Monkey Plasminogen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of monkey Plasminogen in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samp 1 * 96 Tests
Catalog # ANTIA326834-96
Supplier:  ANTIBODIES.COM