ContactUsLinkComponent
Mouse Bile Acid Receptor NR1H4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of mouse Bile Acid Receptor NR1H4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5721-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Bile Acid Receptor NR1H4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of mouse Bile Acid Receptor NR1H4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5721-48
Supplier:  ANTIBODIES.COM