ContactUsLinkComponent
Mouse beta Glucuronidase/GUSB ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse beta Glucuronidase/GUSB in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3860-96
Supplier:  ANTIBODIES.COM
New Product
Mouse beta Glucuronidase/GUSB ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse beta Glucuronidase/GUSB in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3860-96
Supplier:  ANTIBODIES.COM