ContactUsLinkComponent
Human IGFBP5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IGFBP5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 96 Tests
Catalog # ANTIA4717-96
Supplier:  ANTIBODIES.COM
New Product
Human IGFBP5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IGFBP5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 96 Tests
Catalog # ANTIA4717-96
Supplier:  ANTIBODIES.COM