ContactUsLinkComponent
Rat Connexin 43/GJA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Connexin 43/GJA1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4387-48
Supplier:  ANTIBODIES.COM
New Product
Rat Connexin 43/GJA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Connexin 43/GJA1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4387-48
Supplier:  ANTIBODIES.COM