ContactUsLinkComponent
Human Cripto1/CRIPTO ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Cripto1/CRIPTO in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA4809-96
New Product
Human Cripto1/CRIPTO ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Cripto1/CRIPTO in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA4809-96