ContactUsLinkComponent
Human Integrin beta 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Integrin beta 1 in tissue homogenates, cell lysates, cell culture supernates or other biological f 1 * 48 Tests
Catalog # ANTIA5109-48
Supplier:  ANTIBODIES.COM
New Product
Human Integrin beta 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Integrin beta 1 in tissue homogenates, cell lysates, cell culture supernates or other biological f 1 * 48 Tests
Catalog # ANTIA5109-48
Supplier:  ANTIBODIES.COM