ContactUsLinkComponent
Porcine Caveolin-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of porcine Caveolin-1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 96 Tests
Catalog # ANTIA4103-96
Supplier:  ANTIBODIES.COM
New Product
Porcine Caveolin-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of porcine Caveolin-1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 96 Tests
Catalog # ANTIA4103-96
Supplier:  ANTIBODIES.COM