ContactUsLinkComponent
Mouse Connexin 43/GJA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse Connexin 43/GJA1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4385-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Connexin 43/GJA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse Connexin 43/GJA1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4385-96
Supplier:  ANTIBODIES.COM