ContactUsLinkComponent
Monkey Galectin 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of monkey Galectin 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 96 Tests
Catalog # ANTIA3902-96
Supplier:  ANTIBODIES.COM
New Product
Monkey Galectin 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of monkey Galectin 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 96 Tests
Catalog # ANTIA3902-96
Supplier:  ANTIBODIES.COM