ContactUsLinkComponent
Porcine VEGFA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of porcine VEGFA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA3716-48
Supplier:  ANTIBODIES.COM
New Product
Porcine VEGFA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of porcine VEGFA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA3716-48
Supplier:  ANTIBODIES.COM