ContactUsLinkComponent
Rat TGF beta 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat TGF beta 3 in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4114-48
Supplier:  ANTIBODIES.COM
New Product
Rat TGF beta 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat TGF beta 3 in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4114-48
Supplier:  ANTIBODIES.COM