ATTO 610 DBCO. ATTO 610 IS A CARBOPYRONIN-BASED FLUORESCENT DYE KNOWN FOR ITS STRONG ABSORPTION, HIGH FLUORESCENCE QUANTUM YIELD, AND EXCEPTIONAL PHOTOSTABILITY AND THERMAL STABILITY. 1 * 1 mg
Catalog # AATB70255
Fournisseur: AAT Bioquest