ContactUsLinkComponent
XFD660 AZIDE. XFD660, MANUFACTURED BY AAT BIOQUEST, IS A FAR-RED FLUORESCENT DYE STRUCTURALLY EQUIVALENT TO ALEXA FLUOR™ 660 (THERMOFISHER). 1 * 1 mg
Catalog # AATB70094
Supplier:  AAT Bioquest
New Product
XFD660 AZIDE. XFD660, MANUFACTURED BY AAT BIOQUEST, IS A FAR-RED FLUORESCENT DYE STRUCTURALLY EQUIVALENT TO ALEXA FLUOR™ 660 (THERMOFISHER). 1 * 1 mg
Catalog # AATB70094
Supplier:  AAT Bioquest