ContactUsLinkComponent
Mouse Kidney Androgen-Regulated Protein/KAP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the i n vitro quantitative determination of mouse Kidney Androgen-Regulated Protein/KAP in serum, plasma, tissue homogenate 1 * 96 Tests
Catalog # ANTIA79487-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Kidney Androgen-Regulated Protein/KAP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the i n vitro quantitative determination of mouse Kidney Androgen-Regulated Protein/KAP in serum, plasma, tissue homogenate 1 * 96 Tests
Catalog # ANTIA79487-96
Supplier:  ANTIBODIES.COM