ContactUsLinkComponent
Human CD200 Receptor 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human CD200 Receptor 2 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6555-96
Supplier:  ANTIBODIES.COM
New Product
Human CD200 Receptor 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human CD200 Receptor 2 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6555-96
Supplier:  ANTIBODIES.COM