ContactUsLinkComponent
Human Scavenging Receptor SR-BI ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Scavenging Receptor SR-BI in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6045-48
Supplier:  ANTIBODIES.COM
New Product
Human Scavenging Receptor SR-BI ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Scavenging Receptor SR-BI in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6045-48
Supplier:  ANTIBODIES.COM