ContactUsLinkComponent
Human Nociceptin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Nociceptin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA7865-96
Supplier:  ANTIBODIES.COM
New Product
Human Nociceptin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Nociceptin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA7865-96
Supplier:  ANTIBODIES.COM