ContactUsLinkComponent
Horse Interferon gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of horse Interferon gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 96 Tests
  ANTIA7304-96
New Product
Horse Interferon gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of horse Interferon gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 96 Tests
  ANTIA7304-96