ContactUsLinkComponent
Human Parotid Secretory Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Parotid Secretory Protein in serum, plasma, tissue homogenates or other biological fluid 1 * 96 Tests
Catalog # ANTIA7230-96
Supplier:  ANTIBODIES.COM
New Product
Human Parotid Secretory Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Parotid Secretory Protein in serum, plasma, tissue homogenates or other biological fluid 1 * 96 Tests
Catalog # ANTIA7230-96
Supplier:  ANTIBODIES.COM