ContactUsLinkComponent
Human Granzyme A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Granzyme A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA7714-96
Supplier:  ANTIBODIES.COM
New Product
Human Granzyme A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Granzyme A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA7714-96
Supplier:  ANTIBODIES.COM