ContactUsLinkComponent
Canine Insulin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determin ation of canine Insulin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologic 1 * 96 Tests
Catalog # ANTIA82295-96
Supplier:  ANTIBODIES.COM
New Product
Canine Insulin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determin ation of canine Insulin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologic 1 * 96 Tests
Catalog # ANTIA82295-96
Supplier:  ANTIBODIES.COM